missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Disposable Columns, 2 mL

Empty drip columns (0.5 to 5 mL) with matching frits and caps for packing beaded affinity or desalting resins to use in gravity-flow purification techniques.

Brand:  Thermo Scientific™ 29920

262.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10066073

  • £271.00 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Disposable Plastic Columns are empty columns with matching frits and caps for use in a variety of resin-based affinity or desalting techniques.

Additional Information and Online Ordering

These bulk packages of empty columns are economical and convenient devices for packing affinity purification supports and desalting resins for gravity-flow procedures. Three columns sizes are offered, suitable for procedures requiring 0.5 to 10mL resin-bed volumes. The plastic columns are supplied with matching pairs of polyethylene discs and polypropylene caps (top and bottom). The columns are economically-priced for single use and sufficiently durable to prepare and store columns for multiple uses.

Highlights:

  • For bench top gravity-flow affinity purification and size exclusion chromatography
  • Supplied complete with porous polyethylene discs, stoppers and end caps
  • Columns can be pre-packed and stored until needed
  • Compatible with most types of aqueous buffer eluents commonly used in chromatography
  • Plastic funnels can be attached to expand the buffer reservoir for increased flow rates

Includes:

Trial pack contains two columns of each available size (six columns total), matching column accessories (discs and caps), and one funnel (column reservoir extender)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2 mL
Store at room temperature.
100 columns
Gravity Flow Column
Disposable Columns, 2mL
Pierce™
Spin Column
Empty Column, Polyethylene Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZK388084 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number29920
Lot NumberZJ387470 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog Number29920
Lot NumberZH387469 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number29920
Lot NumberZF387468 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number29920
Lot NumberZC385152 Certificate TypeCertificate of Analysis Date14/04/2024 Catalog Number29920
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Disposable Columns, 2 mL > Columns, polystyrene, 2mL; 100 columns

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.