Learn More
Invitrogen™ THE RNA Storage Solution
The Invitrogen RNA Storage Solution is a buffer that provides greater RNA stability than standard 0.1 mM EDTA or TE Buffer.
Brand: Invitrogen™ AM7001
40.83 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
53.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The Invitrogen RNA Storage Solution is a buffer that provides greater RNA stability than standard 0.1 mM EDTA or TE Buffer. This 1 mM sodium citrate, pH 6.4 ± 0.2 buffer is provided in one bottle containing 50 mL.
The last step in every RNA isolation protocol is to resuspend the purified RNA pellet. After painstakingly isolating the RNA, it is crucial that the pellet be suspended and stored in a safe, RNase-free solution. The RNA Storage Solution has two features that minimize base hydrolysis of RNA: a low pH, and sodium citrate, which is an efficient chelating agent. The RNA Storage Solution is compatible with all common RNA applications such as reverse transcription, in vitro transcription, northern analysis, and nuclease protection assays. The RNA Storage Solution is prepared from the purest reagents available, under scrupulously monitored conditions in a state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to guarantee their purity and performance.
Order Info
Shipping Condition: Room temperature

Specifications
Store at -20°C. Shipped at ambient temperature. |
|
Ambion™ | |
50 mL | |
Liquid |
Tube | |
RNA Storage Reagent | |
Room Temperature |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.