Learn More
Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs
Use for the most accurate labeling. Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs contain derivatized CyDye that carries only one reactive group on each dye molecule for accurate labeling of amine groups.
Brand: Cytiva PA23001
Description
- Convenient way to label antibodies, proteins, drugs, and oligonucleotides with CyDye Fluorescent Dyes.
- For use in ELISA, immunoprecipitation, and blotting applications.
- Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labelings.
Order Info
Each kit contains five vials of dye. Each vial is sufficient for labeling 1mg protein or 30nmol amino groups.

Specifications
For the labelling of biological compounds with Cy3 monofunctional dye | |
Cy3 Mono-Reactive Dye Pack | |
7.2 | |
765.95 | |
Cy3 mono-Functional Dye |
Each | |
Five foil packs each containing dried dye to label 1mg of Protein | |
Labeling 1mg Protein or 30nM Amino groups | |
2°C to 8°C, protected from light | |
Red |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Cy3
Your input is important to us. Please complete this form to provide feedback related to the content on this product.