missing translation for 'onlineSavingsMsg'
Learn More
Amiloride hydrochloride dihydrate, Thermo Scientific™ Product Code.: 15424599

Amiloride hydrochloride dihydrate, Thermo Scientific™

Product Code. 15424599
5g
Click to view available options
Quantity:
1g
5g
This item is not returnable. View return policy

Product Code. 15424599

Brand: Thermo Scientific Alfa Aesar J62168.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium channel blocker

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Amiloride hydrochloride dihydrate is used as a calcium channel and sodium channel protein inhibitor. It reduces electrical potential across tubular epithelium, inhibits angiogenesis and inhibits capillary morphogenesis completely and reversibly at approximately 130 μM. Amiloride • HCl is an inhibitor of Sodium/Potassium-ATPase Protein. Defines the I2A-amiloride sensitive and I2B-amiloride insensitive imidazoline binding Blocks TRPP3, acid sensing- (ASIC) and mechanogated membrane-ion channels, as well as the Na+/H+ exchanger. Also inhibits urokinase-type plasminogen activator (uPA); has no effect on tissue-type plasminogen activator.

Solubility
Soluble in water at 50mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17440-83-4
Molecular Formula C6H13Cl2N7O3
Molecular Weight (g/mol) 302.116
MDL Number MFCD00058197
InChI Key LTKVFMLMEYCWMK-UHFFFAOYSA-N
Synonym amiloride hydrochloride dihydrate, modamide, nirulid, amiloride hcl, amiloride hcl dihydrate, amilorid hydrochlorid-2-wasser, unii-fzj37245uc, amiloride hydrochloride usan:usp, amiloride dihydrate hydrochloride, n-amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride dihydrateShow MoreShow Less
PubChem CID 68540
ChEBI CHEBI:2640
IUPAC Name 3,5-diamino-6-chloro-N-(diaminomethylidene)pyrazine-2-carboxamide;dihydrate;hydrochloride
SMILES C1(=C(N=C(C(=N1)Cl)N)N)C(=O)N=C(N)N.O.O.Cl

Specifications

Quantity 5g
UN Number UN2811
Merck Index 14,406
Solubility Information Soluble in water at 50mg/ml
Formula Weight 266.09
Chemical Name or Material Amiloride hydrochloride dihydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW09F024 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog NumberJ62168.06,J62168.03
Lot NumberX29K002 Certificate TypeCertificate of Analysis Date13/11/2024 Catalog NumberJ62168.06,J62168.03
Lot NumberU09K014 Certificate TypeCertificate of Analysis Date14/08/2024 Catalog NumberJ62168.06,J62168.03
Lot NumberP05K032 Certificate TypeCertificate of Analysis Date12/03/2024 Catalog NumberJ62168.06,J62168.03
Lot NumberQ05J018 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberJ62168.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Amiloride hydrochloride dihydrate, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.