missing translation for 'onlineSavingsMsg'
Learn More
(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals Product Code.: 11392277

(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals

Product Code. 11392277
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 11392277

Brand: Thermo Scientific Alfa Aesar L09833.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(Aminomethyl)phosphonic acid is used in the preparation of (acetylamino-methyl)-phosphonic acid by reacting with acetic anhydride. Further, it is used in the development of immobilized metal affinity chromatography column based on organic-inorganic hybrid silica monolith.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Aminomethyl)phosphonic acid is used in the preparation of (acetylamino-methyl)-phosphonic acid by reacting with acetic anhydride. Further, it is used in the development of immobilized metal affinity chromatography column based on organic-inorganic hybrid silica monolith.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-51-9
Molecular Formula CH6NO3P
Molecular Weight (g/mol) 111.037
MDL Number MFCD00008105
InChI Key MGRVRXRGTBOSHW-UHFFFAOYSA-N
Synonym aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethylShow MoreShow Less
PubChem CID 14017
ChEBI CHEBI:28812
IUPAC Name aminomethylphosphonic acid
SMILES C(N)P(=O)(O)O

Specifications

Melting Point ∽290°C (decomposition)
Quantity 1 g
Beilstein 1701219
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 111.04
Percent Purity 99%
Chemical Name or Material (Aminomethyl)phosphonic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251290 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL09833.MD,L09833.06,L09833.03
Lot Number10195772 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL09833.MD,L09833.06,L09833.03
Lot Number10003440 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL09833.MD,L09833.06,L09833.03
Lot Number10237105 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL09833.MD,L09833.06,L09833.03
Lot Number10113305 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL09833.MD,L09833.06,L09833.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.