Learn More
Antimony(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Antimony(III) oxide finds use as flame retardant wherein it synergistically acts with halogenated materials. It is employed in opacifying glasses, ceramics and enamels. In the preparation of optical and ruby glass, antimony trioxide serves as a bubble remover. Further, it is a useful catalyst in the production of polyethylene terephthalate (PET plastic) and the vulcanization of rubber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1550°C (sublimation) |
Melting Point | 656°C |
Color | White |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99.99% (Metals basis) |
Solubility Information | Very slightly soluble in water; soluble in potassium hydroxide,hydrochloricacid and acetic acid; insoluble in organic solvents. |
Formula Weight | 291.5 |
Odor | Odorless |
Density | 5.67 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X15K037 | Certificate of Analysis | 13/11/2024 | 010641.06,010641.18,010641.22,010641.14 |
R02K016 | Certificate of Analysis | 14/05/2024 | 010641.06,010641.18,010641.22,010641.14 |
S07J059 | Certificate of Analysis | 23/04/2024 | 010641.06,010641.18,010641.22,010641.14 |
Y23I027 | Certificate of Analysis | 05/01/2023 | 010641.06,010641.18,010641.22,010641.14 |
P25H039 | Certificate of Analysis | 29/07/2022 | 010641.06,010641.18,010641.22,010641.14 |
Safety and Handling

- Antimony(III) oxide
- Warning
- Carcinogenicity Category 2
- H351-Suspected of causing cancer.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Antimony trioxide, Lead monoxide, Arsenic trioxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.