Learn More
Karl Fischer Aqualine™ Complete 1, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Single-component reagent; Pyridine free
Brand: Fisher Chemical K/1900/15
213.64 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
View more versions of this product
Description
- Single-component volumetric reagent for high water content analysis
- Water equivalent: 1.0mg H2O/mL
- Unlimited water capacity
- Perform titer determination before use
- Pyridine free

Specifications
Aqualine™ Complete 1 | |
Karl Fischer Reagent | |
1 L | |
1.2mg of water/mL min. | |
Liquid | |
81734 |
Colorless | |
Pyridine free | |
111-90-0, 288-32-4, 7446-09-5, 7553-56-2 | |
Amber glass bottle | |
Solution |
Safety and Handling
- Aqualine Complete 1
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P264-Wash face, hands and anyA§ thoroughly after handling.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.