missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AR-M 1000390 hydrochloride, Tocris Bioscience™
Low-internalizing δ agonist
£182.60 - £683.92
Chemical Identifiers
CAS | 209808-47-9 |
---|---|
Molecular Formula | C23H29ClN2O |
Molecular Weight (g/mol) | 384.948 |
InChI Key | OTXTZCLQEGSAMW-UHFFFAOYSA-N |
Synonym | ar-m 1000390 hydrochloride, arm390 hydrochloride, arm-390hcl, n,n-diethyl-4-phenyl-4-piperidinylidenemethyl-benzamide hydrochloride, n,n-diethyl-4-phenyl piperidin-4-ylidene methyl benzamide hydrochloride, arm1000390 hydrochloride, n,n-diethyl-4-phenyl-4-piperidinylidenemethyl benzamide hydrochloride |
PubChem CID | 76848958 |
IUPAC Name | N,N-diethyl-4-[phenyl(piperidin-4-ylidene)methyl]benzamide;hydrochloride |
SMILES | CCN(CC)C(=O)C1=CC=C(C=C1)C(=C2CCNCC2)C3=CC=CC=C3.Cl |
Description
Non-peptidic, low-internalizing δ-selective opioid receptor agonist; derivative of SNC 80 (Cat. No. 0764). Does not trigger acute desensitization of the analgesic response; reduces CFA-induced hyperalgesia. Brain penetrant following systemic administration.Chemical Identifiers
209808-47-9 | |
384.948 | |
ar-m 1000390 hydrochloride, arm390 hydrochloride, arm-390hcl, n,n-diethyl-4-phenyl-4-piperidinylidenemethyl-benzamide hydrochloride, n,n-diethyl-4-phenyl piperidin-4-ylidene methyl benzamide hydrochloride, arm1000390 hydrochloride, n,n-diethyl-4-phenyl-4-piperidinylidenemethyl benzamide hydrochloride | |
N,N-diethyl-4-[phenyl(piperidin-4-ylidene)methyl]benzamide;hydrochloride |
C23H29ClN2O | |
OTXTZCLQEGSAMW-UHFFFAOYSA-N | |
76848958 | |
CCN(CC)C(=O)C1=CC=C(C=C1)C(=C2CCNCC2)C3=CC=CC=C3.Cl |
Specifications
209808-47-9 | |
ar-m 1000390 hydrochloride, arm390 hydrochloride, arm-390hcl, n,n-diethyl-4-phenyl-4-piperidinylidenemethyl-benzamide hydrochloride, n,n-diethyl-4-phenyl piperidin-4-ylidene methyl benzamide hydrochloride, arm1000390 hydrochloride, n,n-diethyl-4-phenyl-4-piperidinylidenemethyl benzamide hydrochloride | |
CCN(CC)C(=O)C1=CC=C(C=C1)C(=C2CCNCC2)C3=CC=CC=C3.Cl | |
384.948 | |
384.94 | |
AR-M 1000390 hydrochloride |
C23H29ClN2O | |
OTXTZCLQEGSAMW-UHFFFAOYSA-N | |
N,N-diethyl-4-[phenyl(piperidin-4-ylidene)methyl]benzamide;hydrochloride | |
76848958 | |
>97% |
Safety and Handling
Recommended Storage : Store at -20°C