missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Microplate Sealing Film and Tapes
GREENER_CHOICE

Offer great economy for disposable ease.

£35.72 - £2460.00


Products 20
Product Code Brand Material Product Type Dimensions (L x W) Sterility Price Quantity & Availability  
Product Code Brand Material Product Type Dimensions (L x W) Sterility Price Quantity & Availability  
12732796
View Documents
Axygen™
BF-400
Rayon Breathable Sealing Film 153 x 82.3 mm Non-sterile N/A N/A N/A
11326254
View Documents Promotion Details
Axygen™
BF-400
Rayon Breathable Sealing Film 153 x 82.3 mm Non-sterile
£156.00
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
11336254
View Documents
Axygen™
BF-400-S
Rayon Breathable Sealing Film 153 x 82.3 mm Sterile N/A N/A N/A
12742796
View Documents
Axygen™
BF-400-S
Rayon Breathable Sealing Film 153 x 82.3 mm Sterile
£631.53
Pack of 500
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
 
12623616
GREENER_CHOICE
View Documents
Axygen™
HSF-AS-300-L
Aluminum Sealing Film Roll 115 x 500 m Non-sterile N/A N/A N/A
12633726
View Documents
Axygen™
HSF-AS-300-P
Aluminum Sealing Film Roll 78 x 610 m Non-sterile N/A N/A N/A
12613616
GREENER_CHOICE
Vedere i documenti
Axygen™
HSF-TS-L
Polyester CyclerSeal Sealing Film Roll 115 x 500 m Non-sterile N/A N/A N/A
12645926
Vedere i documenti
Axygen™
HSF-TS-P
Polyester CyclerSeal Sealing Film Roll 78 x 610 m Non-sterile N/A N/A N/A
12557947
GREENER_CHOICE
Vedere i documenti
Axygen™
MF-300
Litto or Ethylene or Vinyl Heat Seal Film 127.2 x 79.5 mm Non-sterile N/A N/A N/A
12752796
Vedere i documenti
Axygen™
PCR-AS-200
Aluminum Sealing Film 146 x 79.2 mm Non-sterile N/A N/A N/A
12577947
Vedere i documenti Dettagli della promozione
Axygen™
PCR-AS-200
Aluminum Sealing Film 146 x 79.2 mm Non-sterile
£197.00
Pack of 100
In Stock
to see stock.
Edge
Aggiungi al carrello
 
12762796
Vedere i documenti
Axygen™
PCR-AS-600
Aluminum Sealing Film 133.3 x 79.2 mm Non-sterile N/A N/A N/A
11380215
Vedere i documenti
Axygen™
PCR-SP-S
Polyester AxySeal Sealing Film 146 x 79.6 mm Sterile N/A N/A N/A
12782796
Vedere i documenti
Axygen™
PCR-SP-S
Polyester AxySeal Sealing Film 146 x 79.6 mm Sterile
£326.34
Pack of 500
In Stock
to see stock.
Aggiungi al carrello
 
12772796
Vedere i documenti Dettagli della promozione
Axygen™
PCR-SP
Polyester AxySeal Sealing Film 146 x 79.6 mm Non-sterile
£369.00
Pack of 500
In Stock
to see stock.
Edge
Aggiungi al carrello
 
11336074
Vedere i documenti
Axygen™
PCR-SP
Polyester AxySeal Sealing Film 146 x 79.6 mm Non-sterile N/A N/A N/A
12014538
Vedere i documenti
Axygen™
PCR-TS
Polyester CyclerSeal Sealing Film 135.5 x 80 mm Non-sterile N/A N/A N/A
13131824
Vedere i documenti Dettagli della promozione
Axygen™
PCR-TS
Polyester CyclerSeal Sealing Film 135.5 x 80 mm Non-sterile
£396.00
Pack of 500
In Stock
to see stock.
Aggiungi al carrello
 
11304325
Vedere i documenti Dettagli della promozione
Axygen™
UC-500
Polyolefin UltraClear Sealing Film Not Available Non-sterile
£325.03
Pack of 100
In Stock
to see stock.
Aggiungi al carrello
 
13191604
Vedere i documenti
Axygen™
UC-500
Polyolefin UltraClear Sealing Film Not Available Non-sterile N/A N/A N/A
Descrizione

Descrizione

For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. Sealing film temperature ranges from -40°C to +104°C.

Breathable

  • Allow effective gas exchange for cellular and bacterial cultivation — while preventing contamination
  • Seal polypropylene and polystyrene culture plates, 96- and 384-well plates including other assay plates
Aluminum
  • Provides exceptional seal on multi-well plates with varied temperatures and reagents
  • With compression mats, films and tapes seal during thermocycling
  • Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
  • Pierceable, PCR-AS-200
CyclerSeal
  • Exceptional seal across entire plate safeguards samples during transport or storage
  • When used with Axygen's compression mat, this polyester film eliminates well-to-well contamination and cross-over in PCR applications
AxySeal
  • Polyester-based with uniformly and consistently applied acrylic adhesive to eliminate the edge effect in sensitive ELISA assays
  • Suitable for sealing tissue culture plates, short-term storage and incubation, transport and the containment of biohazardous solutions
  • Available in both nonsterile and sterile packaging

Videos
SDS
Documenti

Documenti

Product Certifications
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
Axygen™ Microplate Sealing Film and Tapes

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato