missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates
GREENER_CHOICEBIOPHARMA

For sample collection, in-vitro growth chambers and long term storage.

Brand:  Axygen™ P-DW-10ML-24-C

211.02 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.


 View more versions of this product

Product Code. 12537837

  • £287.00 / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

Used for sample collection, in vitro growth chambers and long-term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • Excellent chemical resistance
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Centrifugation to 4,000 RCF
  • Diamond shaped wells allow complete sample recovery
  • Black and white plates are available by special order
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    Standard
    Autoclavable
    Polypropylene
    24
    Square
    4000xG
    5/Pk.
    -80°C
    4 x 6
    Clear
    10,000 μL
    Non-sterile
    Sample collection, in-vitro growth chambers and long term storage
    25/Cs.
    -112°F
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents Certificates

    Certificates

    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot Number34524017 Certificate TypeCertificate of Quality Date10/04/2025 Catalog NumberP-DW-10ML-24-C
    Lot Number01525128 Certificate TypeCertificate of Quality Date10/04/2025 Catalog NumberP-DW-10ML-24-C
    Lot Number32424002 Certificate TypeCertificate of Quality Date01/04/2025 Catalog NumberP-DW-10ML-24-C
    Lot Number22524007 Certificate TypeCertificate of Quality Date13/02/2025 Catalog NumberP-DW-10ML-24-C
    Lot Number21624016 Certificate TypeCertificate of Quality Date11/02/2025 Catalog NumberP-DW-10ML-24-C
    Special Offers

    Special Offers

    • Multiple Manufacturers
      Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
      Promo Code: 24155

      Expires2025-04-30

      Days

      19

      Hours

      5

      Minutes

      33

      Seconds

      9
      Hot Deal

      *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

      Terms & Conditions See Full Promo Details
    • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
      Corning Lab Products Offer
      Promo Code24073

      Expires2025-04-25

    • Multiple Manufacturers
      Master liquid handling with ease.
      Promo Code: 24122

      Expires2025-04-13

      Days

      2

      Hours

      5

      Minutes

      33

      Seconds

      9
    • Multiple Manufacturers
      Save up to 94% on Teaching Lab Essentials
      Promo Code25721

      Expires2025-05-01

      New Offer
      Save Now
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Axygen™ Storage Microplates > 24-well; 10 mL; No; Square

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.