missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Clear Standard Modules

Perform applications that deliver well-to-well reproducibility with these immuno clear standard modules. Manufactured to fit standard equipment.

Brand:  Thermo Scientific™ 441653

336.99 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10574941

  • £592.00 / Pack of 60
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

No
Certified for adsorption of IgG; CV of <5% between wells
Standard
Colorimetric
60 Cases
Frame
380 μL
250 μL
Round, C Bottom
10
60/Cs.
C8
Hydrophilic
Yes
Without Lid
8 x 12
Clear
Polystyrene
96
MaxiSorp
60
None
Antibodies (Glycoproteins)
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number177454 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number441653
Lot Number165566 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number441653
Lot Number180557 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number441653
Lot Number170086 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number441653
Lot Number181972 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number441653
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Immuno Clear Standard Modules > Nunc; C8 StarWell; MaxiSorp; 380μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.