missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Clear Standard Modules
BIOPHARMA

Perform applications that deliver well-to-well reproducibility with these immuno clear standard modules. Manufactured to fit standard equipment.

Brand:  Thermo Scientific™ 475078

363.99 GBP valid until 2025-12-31
Use promo code "24284" to get your promotional price.


 View more versions of this product

Product Code. 10386231

  • £436.00 / Pack of 60
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

No
Certified for adsorption of IgG; CV of <5% between wells
Strip Plate
Colorimetric
60 Cases
Clear
Polystyrene
250 μL
Round
10
60/Cs.
U8
Hydrophilic
Yes
Without Lid
8 x 12
Adsorption
Strip Plate
96
MaxiSorp
60
None
Antibodies (Glycoproteins)
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number185276 Certificate TypeCertificate of Compliance Date09/01/2025 Catalog Number475078
Lot Number169403 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number475078
Lot Number171580 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number475078
Lot Number176685 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number475078
Lot Number166076 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number475078
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Immuno Clear Standard Modules > Nunc; U8; MaxiSorp; 300μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.