missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Clear Standard Modules
GREENER_CHOICEBIOPHARMA

Perform applications that deliver well-to-well reproducibility with these immuno clear standard modules. Manufactured to fit standard equipment.

£161.00 - £1390.00

Specifications

Detection Method Colorimetric
Color Clear
Plate Blocking None
View More Specs

Products 25
Product Code Brand Surface Treatment Volume (Metric) Working Well Shape Well Design Price Quantity & Availability  
Product Code Brand Surface Treatment Volume (Metric) Working Well Shape Well Design Price Quantity & Availability  
10721634
GREENER_CHOICE
View Documents
Thermo Scientific™
434797
MaxiSorp 350 μL Round, Flat Bottom (F-well) F8
£1390.00
Pack of 120
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
 
10294021
View Documents
Thermo Scientific™
441254
PolySorp 250 μL Curved Edges, Flat Bottom (C-well) C8 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10574941
View accessory products View Documents
Thermo Scientific™
441653
MaxiSorp 250 μL Round, C Bottom C8
£592.00
Pack of 60
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
 
10274071
View accessory products View Documents
Thermo Scientific™
444865
PolySorp 250 μL Round, C Bottom C8
£433.00
Pack of 60
In Stock
to see stock.
Add to basket
 
10172881
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
445101
MaxiSorp 250 μL Round, C Bottom C8
£433.00
Pack of 60
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
 
10032391
View accessory products View Documents
Thermo Scientific™
467120
MediSorp 350 μL Round, Flat Bottom (F-well) F8
£1125.00
Pack of 60
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
 
10052411
View accessory products View Documents
Thermo Scientific™
467140
MultiSorp 350 μL Round, Flat Bottom (F-well) F8
£444.00
Pack of 60
Please to purchase this item. Need a web account? Register with us today!
10797921
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
467466
MaxiSorp 350 μL Round, Flat Bottom (F-well) F16 (2x8 wells)
£303.00
Pack of 60
In Stock
to see stock.
Add to basket
 
10554831
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
468667
MaxiSorp 350 μL Round, Flat Bottom (F-well) F8
£1110.00
Pack of 60
In Stock
to see stock.
Edge
Add to basket
 
10673832
View accessory products View Documents
Thermo Scientific™
469078
PolySorp 350 μL Round, Flat Bottom (F-well) F8
£433.00
Pack of 60
In Stock
to see stock.
Add to basket
 
12799995
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
469264
Hydrophilic 250 μL Round U16
£179.00
Pack of 320
In Stock
to see stock.
Add to basket
 
10174811
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
469914
MaxiSorp 350 μL Round, Flat Bottom (F-well) F16 (2x8 wells)
£447.00
Pack of 320
In Stock
to see stock.
Add to basket
 
10738091
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
469922
PolySorp 350 μL Round, Flat Bottom (F-well) F16 (2x8 wells)
£180.00
Pack of 320
Estimated Shipment: 24-06-2025
to see stock.
Add to basket
 
10227541
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
469949
MaxiSorp 350 μL Round, Flat Bottom (F-well) F8
£466.00
Pack of 640
In Stock
to see stock.
Add to basket
 
10748091
View accessory products View Documents
Thermo Scientific™
469957
PolySorp 350 μL Round, Flat Bottom (F-well) F8
£309.92
Pack of 640
In Stock
to see stock.
Add to basket
 
10244551
View accessory products View Documents
Thermo Scientific™
473709
MaxiSorp 250 μL Round, C Bottom C12
£365.00
Pack of 60
In Stock
to see stock.
Add to basket
 
10505411
View accessory products View Documents
Thermo Scientific™
473717
PolySorp 250 μL Round, C Bottom C12 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10386231
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
475078
MaxiSorp 250 μL Round U8
£436.00
Pack of 60
In Stock
to see stock.
Add to basket
 
10396231
View accessory products View Documents
Thermo Scientific™
475086
PolySorp 250 μL Round U8
£436.00
Pack of 60
In Stock
to see stock.
Add to basket
 
10079074
GREENER_CHOICE
View Documents
Thermo Scientific™
6302
Immulon 2 HB 330 μL Round, Flat Bottom (F-well) F12
£171.00
Pack of 320
In Stock
to see stock.
Add to basket
 
10406165
GREENER_CHOICE
View Documents
Thermo Scientific™
6309
Immulon 2 HB 330 μL Round, Flat Bottom (F-well) F12
£557.00
Pack of 100
In Stock
to see stock.
Add to basket
 
10364025
GREENER_CHOICE
View Documents
Thermo Scientific™
6310
Immulon 1 B 330 μL Round, Flat Bottom (F-well) F12
£537.00
Pack of 100
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
 
10753886
GREENER_CHOICE
View Documents
Thermo Scientific™
6405
Immulon 4 HBX 330 μL Round, Flat Bottom (F-well) F12
£597.00
Pack of 100
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
 
10695757
GREENER_CHOICE
View Documents
Thermo Scientific™
95029100
Enhanced Binding 350 μL Round, Flat Bottom (F-well) F8
£326.00
Pack of 50
In Stock
to see stock.
Add to basket
 
10497305
GREENER_CHOICE
View Documents
Thermo Scientific™
6301
Immulon 1 B 330 μL Round, Flat Bottom (F-well) F12
£161.00
Pack of 320
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
 
Description
Show More
Specifications

Specifications

Colorimetric
Clear
None
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Immuno Clear Standard Modules

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.