missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Clear Standard Modules
BIOPHARMA

Perform applications that deliver well-to-well reproducibility with these immuno clear standard modules. Manufactured to fit standard equipment.

Brand:  Thermo Scientific™ 469949

238.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10227541

  • £466.00 / Pack of 640
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Breakable Strips
Certified for adsorption of IgG; CV of <5% between wells
Standard
Colorimetric
640 Cases
Microplate Strip
8
MaxiSorp
640
None
Antibodies (Glycoproteins)
Hydrophilic
Yes
Without Lid
1 x 8
Clear
Polystyrene
350 μL
Round, Flat Bottom (F-well)
160
640/Cs.
F8
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number185466 Certificate TypeCertificate of Compliance Date20/03/2025 Catalog Number469949
Lot Number185290 Certificate TypeCertificate of Compliance Date14/03/2025 Catalog Number469949
Lot Number184867 Certificate TypeCertificate of Compliance Date21/01/2025 Catalog Number469949
Lot Number182838 Certificate TypeCertificate of Compliance Date04/12/2024 Catalog Number469949
Lot Number170079 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number469949
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Immuno Clear Standard Modules > Nunc; F8; MaxiSorp; 400μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.