Learn More
Axygen™ Storage Microplates



For sample collection, in-vitro growth chambers and long term storage.
Brand: Axygen™ P-DW-11-C
154.50 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Used for sample collection, in vitro growth chambers and long-term storage.
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
Specifications
ANSI, SLAS 1-2004 | |
Without Lid | |
Autoclavable | |
Polypropylene | |
96 | |
Round | |
4000xG | |
5/Pk. | |
-80°C |
Standard | |
8 x 12 | |
Clear | |
1.1 mL | |
Non-sterile | |
Sample collection, in-vitro growth chambers and long term storage | |
50/Cs. | |
-112°F |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
22924009 | Certificate of Quality | 12/04/2025 | P-DW-11-C |
25624051 | Certificate of Quality | 27/03/2025 | P-DW-11-C |
25624050 | Certificate of Quality | 27/03/2025 | P-DW-11-C |
22724013 | Certificate of Quality | 11/03/2025 | P-DW-11-C |
21524014 | Certificate of Quality | 04/03/2025 | P-DW-11-C |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.