missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azepan-2-one oxime, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS | 19214-08-5 |
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C6H12N2O |
MDL Number | MFCD00053062 |
Quantity | 1 g |
Synonym | azepan-2-one oxime, n-azepan-2-ylidene hydroxylamine, 2z-azepan-2-one oxime, 2h-azepin-7-amine,3,4,5,6-tetrahydro-n-hydroxy, n-2e-azepan-2-ylidene hydroxylamine, 2-hydroxyimino azaperhydroepine, n-2z-azepan-2-ylidene hydroxylamine, caprolactam-oxime, hieh@hpdeewafjjadp, azepan-2-oneoxime |
InChI Key | PPOHPUOKXMNCCI-UHFFFAOYSA-N |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Azepan-2-one oxime, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction