missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ LDPE Specimen Container with Hinged Cap
Unbreakable and lightweight specimen containers. AzlonTM LDPE specimen containers with hinged caps are perfect for many laboratory applications.
£6.34 - £22.95
Product Code | Brand | Diameter (Metric) | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | |||||
10717465
|
Azlon
BGG308 |
28 mm | 10 mL | 32 mm |
£18.65
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10584745
|
Azlon
BGG311 |
32 mm | 20 mL | 36 mm | N/A | ||||||
12783335
|
Azlon
BGG304 |
12 mm | 2.5 mL | 40 mm | N/A | ||||||
12793335
|
Azlon
BGG312 |
25 mm | 25 mL | 78 mm |
£20.45
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10739555
|
Azlon
BGG306 |
15 mm | 5 mL | 49 mm |
£17.00
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10135874
|
Azlon
BGG302 |
8 mm | 1 mL | 38 mm | N/A | ||||||
Description
- Containers with captive hinged cap suitable for many applications
- Lightweight and unbreakable in normal use
- Can withstand temperatures up to 80°C
Specifications
Specimen Container | |
Yes | |
LDPE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Azlon™ LDPE Specimen Container with Hinged Cap