missing translation for 'onlineSavingsMsg'
Learn More
Barium hydroxide monohydrate, 95%, Thermo Scientific Chemicals Product Code.: 15412537

Barium hydroxide monohydrate, 95%, Thermo Scientific Chemicals

Product Code. 15412537
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 15412537

Brand: Thermo Scientific Alfa Aesar 045888.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Barium hydroxide monohydrate is used in water purification. It is used to prepare lubricating and oil additives as well as a precursor to other barium compounds. It is also used to dehydrate and remove sulfate from various products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 22326-55-2
Molecular Formula BaH4O3
Molecular Weight (g/mol) 189.356
MDL Number MFCD00149151
InChI Key GKQTUHKAQKWLIN-UHFFFAOYSA-L
Synonym barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a tShow MoreShow Less
PubChem CID 16211219
IUPAC Name barium(2+);dihydroxide;hydrate
SMILES O.[OH-].[OH-].[Ba+2]

Spezifikation

Color White
Physical Form Crystalline
Quantity 100 g
Linear Formula Ba(OH)2·H2O
UN Number UN3262
Solubility Information Soluble in water.
Formula Weight 189.38 (171.36 Anhydrous)
Percent Purity 95%
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 3.743 g/mL
Chemical Name or Material Barium hydroxide monohydrate
Mehr anzeigen Weniger anzeigen

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN05L007 Certificate TypeCertificate of Analysis Date07/02/2025 Catalog Number045888.36,045888.22
Lot NumberZ06K010 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog Number045888.36,045888.22
Lot NumberS28K038 Certificate TypeCertificate of Analysis Date25/07/2024 Catalog Number045888.36,045888.22
Lot NumberX26J026 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number045888.36,045888.22
Lot NumberU15J086 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number045888.36,045888.22
compliance-icons compliance-icons
Product Identifier
  • Barium hydroxide monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
  • P363-Wash contaminated clothing before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Barium hydroxide, monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium hydroxide monohydrate, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.