Learn More
Barium hydroxide monohydrate, 95%, Thermo Scientific Chemicals
Description
Barium hydroxide monohydrate is used in water purification. It is used to prepare lubricating and oil additives as well as a precursor to other barium compounds. It is also used to dehydrate and remove sulfate from various products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 500 g |
Linear Formula | Ba(OH)2·H2O |
UN Number | UN3262 |
Solubility Information | Soluble in water. |
Formula Weight | 189.38 (171.36 Anhydrous) |
Percent Purity | 95% |
Odor | Odorless |
Sensitivity | Air sensitive; Hygroscopic |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N05L007 | Certificate of Analysis | 07/02/2025 | 045888.36,045888.22 |
Z06K010 | Certificate of Analysis | 11/12/2024 | 045888.36,045888.22 |
S28K038 | Certificate of Analysis | 25/07/2024 | 045888.36,045888.22 |
X26J026 | Certificate of Analysis | 03/11/2023 | 045888.36,045888.22 |
U15J086 | Certificate of Analysis | 24/10/2023 | 045888.36,045888.22 |
Safety and Handling


- Barium hydroxide monohydrate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- EUH071-Corrosive to the respiratory tract.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Barium hydroxide, monohydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.