missing translation for 'onlineSavingsMsg'
Learn More
Barium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11380849

Barium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11380849
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11380849

Brand: Thermo Scientific Alfa Aesar 010648.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Barium nitrate is used by the oil and gas industries to make drilling muds. It is used to make  paints, bricks, tiles, glass, rubber, and other barium compounds. It also used in is used in signal flares and water softener. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Synonym barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2Show MoreShow Less
PubChem CID 24798
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 592°C (decomposition)
Color White
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water. Slightly soluble in ethanol and acetone. Partially soluble in cold water.
IUPAC Name barium(2+);dinitrate
Formula Weight 261.35
Sensitivity Hygroscopic
Appearance White crystalline
Density 3.24
Chemical Name or Material Barium nitrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW23K008 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog Number010648.14,010648.36,010648.22,010648.06
Lot NumberW21K006 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog Number010648.14,010648.36,010648.22,010648.06
Lot NumberS20K032 Certificate TypeCertificate of Analysis Date27/06/2024 Catalog Number010648.14,010648.36,010648.22,010648.06
Lot NumberM30G018 Certificate TypeCertificate of Analysis Date30/06/2022 Catalog Number010648.14,010648.36,010648.22,010648.06
Lot NumberM09G021 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010648.14,010648.36,010648.22,010648.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.