missing translation for 'onlineSavingsMsg'
Learn More

Barium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 15495955
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
10022-31-8
BaN2O6
261.34
MFCD00003442
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
24798
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
This item is not returnable. View return policy
10022-31-8
BaN2O6
261.34
MFCD00003442
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
24798
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Barium nitrate is used by the oil and gas industries to make drilling muds. It is used to make  paints, bricks, tiles, glass, rubber, and other barium compounds. It also used in is used in signal flares and water softener. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.99% (Metal basis)
IUPAC Name barium(2+);dinitrate
Formula Weight 261.35
Appearance White crystalline
Density 3.24g/cm3
Chemical Name or Material Barium nitrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'