Learn More
Cytiva BAS Storage Phosphor Screens
For Neutron Detection, Size 20 x 40 cm
Brand:  Cytiva 29-0171-33
Product Code. 15327457
  • £2537.00 / Each
Estimated Shipment: 15-05-2025
Explore more special offers
Add to basket
Description
Description
  • Requires approximately one-tenth the exposure time of traditional autoradiography film
  • Ten to one hundred times more sensitive than film, depending on isotope and sample type
  • Wide linear dynamic range (five orders of magnitude) enables quantitation and visualization of both weak and strong signals in a single exposure
  • Reusable screens do not require chemicals, darkroom, or other special treatment for use
  • Each step can be performed at the lab bench under normal lighting conditions at room temperature
  • Results are digitized using a storage phosphor imaging system (e.g., Typhoon, Storm, or PhosphorImager) and quantitated using ImageQuant software
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 x 40 cm
Neutron
Neutron Detection
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva BAS Storage Phosphor Screens > 20 x 40 cm