missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dry-blend Buffered Packs

Our quality control ensures that every pack of dry-blend buffers yields consistent solutions without weighing or pH adjustment. Simply empty contents of one foil envelope pack into a beaker, add appropriate solvents, and stir to dissolve.

Brand:  Thermo Scientific™ 28384

159.60 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.



 View more versions of this product

Product Code. 10628694

  • £172.00 / Pack of 40

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Store at room temperature.
BupH™
40 Packs
Powder
Dry Blend
Borate Buffer
8.5
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAA402501 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number
Lot NumberZI400316 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number28384
Lot NumberZJ401904 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number28384
Lot NumberZG395746 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number28384
Lot NumberZF388055 Certificate TypeCertificate of Analysis Date12/06/2024 Catalog Number28384
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Dry-blend Buffered Packs > BupH Borate Buffer Packs, 40 Packs

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.