missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetal Bovine Serum Stain Buffer, BD
For the immunofluorescent staining of single-cell suspensions
Brand: BD Biosciences 554656
170.62 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Prepared from lymphoid tissues, bone marrow, peripheral blood,cultured cells
- Maintain cell viability and maximize fluorescence signal intensities by pH-sensitive fluorochromes
- Contains metabolic inhibitor, sodium azide
- For dilution, application of fluorescent reagents and suspension
- Aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
Flow cytometry

Specifications
4°C | |
500mL | |
Staining Buffer |
Heat-inactivated FBS proteins and less-than-or-equal-to 0.09% sodium azide | |
RUO |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fetal Bovine Serum Stain Buffer, BD > 500mL
For research use only.
Spot an opportunity for improvement?Share a Content Correction