Learn More
Invitrogen™ eBioscience™ Flow Cytometry Staining Buffer
A buffered saline solution containing fetal bovine serum and sodium azide (0.09%) as a preservative. Designed for use in immunofluorescent staining protocols of cells in suspension.
Brand: Invitrogen™ 00-4222-26
72.36 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Alert:
Contains Sodium azide
Description
- Designed for use in immunofluorescent staining protocols of cells in suspension
- A buffered saline solution containing fetal bovine serum and sodium azide (0.09%) as a preservative
- This buffer can be used for antibody and cell dilution steps, as well as all the wash steps required for the surface staining and flow cytometric analysis
- Contains animal serum proteins to help minimizing non-specific binding of antibodies

Specifications
600 mL | |
eBioscience™ | |
Staining Buffer |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3051958 | Certificate of Analysis | 31/01/2025 | 00-4222-26 |
2963749 | Certificate of Analysis | 31/01/2025 | 00-4222-26 |
2945611 | Certificate of Analysis | 30/01/2025 | 00-4222-26 |
2312105 | Certificate of Analysis | 29/01/2025 | 00-4222-26 |
2486688 | Certificate of Analysis | 29/01/2025 | 00-4222-26 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.