Learn More
BD Retrievagen Antigen Retrieval Systems
Recommended for zinc-fixed and formalin-fixed paraffin embedded tissue sections that show suboptimal or no reactivity with antibodies for immunohistochemical staining
Brand:  BD 550524
312.46 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15811807
  • £449.00 / 1 L
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
10% neutral buffered formalin fixes the tissue by cross-linking proteins that may cause a conformational change or “masking” of the antigenic epitope leading to reduced or absent antibody reactivity. The BD Retrievagen Antigen Retrieval Systems are designed to unmask the epitope and cause the enhancement of antibody staining.
  • Retrievagen A (pH 6.0) is provided as two separate solutions that need to be mixed
  • Retrievagen B (pH 9.5) is provided as a ready-to-use solution
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25mL Retrievagen A Solution 1 and 100mL Retrievagen A Solution 2
Retrieval Reagent
Immunohistochemical staining
RUO
Take 18 mL of BD Retrievagen A Solution 1 and 82 mL of BD Retrievagen Solution 2 and bring up the volume to 1 L with distilled Water
125 mL
6
Retrievagen A
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Retrievagen Antigen Retrieval Systems > Retrievagen A (pH 6.0)