missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ E-A-R™ E-A-Rsoft Yellow Earplugs
Offers high attenuation and are made from a slow expanding, PU foam material. 3M E-A-R Polyurethane Earplug – 36dB.
Brand: 3M™ 7100111802
This item is not returnable.
View return policy
Description
- Made from a slow expanding, PU foam material
- Provides evenly distributed pressure, giving flexibility, and a good seal with optimum comfort.
- Bright, neon color makes it appealing to wearers

Specifications
Yellow Neon | |
For general industrial use | |
36 dB | |
Ear Plugs | |
Roll-down |
CE Approved | |
250 Pairs | |
Uncorded | |
Polyurethane |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ E-A-R™ E-A-Rsoft Yellow Earplugs > Packaging: 250 pair
Spot an opportunity for improvement?Share a Content Correction