missing translation for 'onlineSavingsMsg'
Learn More

3M™ E-A-R™ E-A-Rsoft Yellow Earplugs

Offers high attenuation and are made from a slow expanding, PU foam material. 3M E-A-R Polyurethane Earplug – 36dB.

Brand:  3M™ 7000052845

52.05 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11779305

  • £98.95 / 200 pairs
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made from a slow expanding, PU foam material
  • Provides evenly distributed pressure, giving flexibility, and a good seal with optimum comfort.
  • Bright, neon color makes it appealing to wearers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Yellow Neon
Vinyl
36 dB
Ear Plugs
Roll-down
CE Approved
200/Pk.
Corded
Polyurethane
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ E-A-R™ E-A-Rsoft Yellow Earplugs > Color: Yellow Neon; Quantity: 200/Pk.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.