Learn More
Cytiva Benchkote Sheet for ÄKTA Systems
Protects the top buffer trays from buffer spillages and salt deposits
Brand: Cytiva 2300-10065
This item is not returnable.
View return policy
Description
- Cover the top buffer tray to protect from buffer spillages and salt deposits
- Simplify daily and weekly preventive maintenance of the top buffer tray of ÄKTA systems
- Manufactured of absorbent, impermeable material that is designed to protect lab surfaces against hazardous spills
- Smooth, absorbent Whatman paper quickly absorbs spills
- Laminated polyethylene layer prevents flow-through to the working surface
- Fits the top buffer tray on ÄKTA go, ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems.
- Convenient and easy to use
Specifications
Absorbent Liner | |
Paper, Laminated Polyethylene | |
ÄKTA Start | |
50/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Cytiva Benchkote Sheet for ÄKTA Systems > ÄKTA Start, 50/Pk
Spot an opportunity for improvement?Share a Content Correction