missing translation for 'onlineSavingsMsg'
Learn More
Benzofurazan, 97%, Thermo Scientific Chemicals Product Code.: 11397636

Benzofurazan, 97%, Thermo Scientific Chemicals

Product Code. 11397636
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11397636

Brand: Thermo Scientific Alfa Aesar L11229.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 273-09-6
Molecular Formula C6H4N2O
Molecular Weight (g/mol) 120.111
MDL Number MFCD00068064
InChI Key AWBOSXFRPFZLOP-UHFFFAOYSA-N
Synonym benzofurazan, benzo c 1,2,5 oxadiazole, benzofurazane, 2,1,3-benzoxadiazol, benzo-2,1,3-oxadiazole, 2-oxa-1,3-diaza-2h-isoindene, 3,5-cyclohexadiene-1,2-dione, dioxime, furazan, pubchem22999, acmc-209gwb
PubChem CID 67501
IUPAC Name 2,1,3-benzoxadiazole
SMILES C1=CC2=NON=C2C=C1

Specifications

Melting Point 52°C to 55°C
Boiling Point 80°C (20 mmHg)
Flash Point 76°C (169°F)
Quantity 1 g
Beilstein 112261
Formula Weight 120.11
Percent Purity 97%
Chemical Name or Material Benzofurazan

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU01A032 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL11229.03,L11229.06
Lot Number10195609 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL11229.03,L11229.06
Lot Number10155370 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL11229.03,L11229.06
Lot NumberS25A016 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL11229.03,L11229.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzofurazan, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.