missing translation for 'onlineSavingsMsg'
Learn More
Benzyl alcohol, Puriss. p.a., ACS reagent, ≥99.0% (GC), Honeywell Riedel-de Haën™ Product Code.: 15641270

Benzyl alcohol, Puriss. p.a., ACS reagent, ≥99.0% (GC), Honeywell Riedel-de Haën™

Product Code. 15641270
500 mL, Glass Bottle
Click to view available options
Quantity:
4 x 25 mL
500 mL
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 15641270

Brand: Honeywell-Riedel-de Haen 13160500ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS reagent,≥99.0% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcoholShow MoreShow Less
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Melting Point -15.4°C
Color Colorless
Density 1.045g/mL at 25°C
Boiling Point 203°C to 205°C
Flash Point 96°C (204.8°F)
Ignition Residue ≤0.005% (as SO4)
Ignition Point 817°F
Packaging Glass Bottle
Linear Formula C6H5CH2OH
Refractive Index n20/D 1.539(lit.)
Quantity 500 mL
UN Number UN3334
Beilstein 878307
Vapor Density 3.7 (vs air)
Formula Weight 108.14g/mol
Vapor Pressure 3.75mmHg (77°C), 13.3mmHg (100°C)
Percent Purity ≥99.0% (GC)
Grade puriss. p.a.
Physical Form Liquid
Chemical Name or Material Benzyl Alcohol, ACS reagent
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : benzyl alcohol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyl alcohol, Puriss. p.a., ACS reagent, ≥99.0% (GC), Honeywell Riedel-de Haën™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.