Learn More
Fisherbrand™ Beta Radiation Shields
Ideal for protection from Beta radiation, Fisherbrand™ radiation shields are supplied in small, medium and large sizes and with a 15cm-deep curved base for use with safety trays, or a 30x30cm flat base for under-the-bench protection.
Brand: Fisherbrand™ CSR-S1O
Description
- Beta – Small Fixed 45° Angle – Flat Base 30 x 45cm, Base 30 x 30cm
- Ideal for protection from Beta radiation
Specifications
Flat Base | |
Small Fixed 45° Angle | |
Beta Radiation Shield | |
455 x 300 x 150 mm |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.