missing translation for 'onlineSavingsMsg'
Learn More
Bismuth(III) nitrate oxide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals Product Code.: 11386138

Bismuth(III) nitrate oxide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Product Code. 11386138
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11386138

Brand: Thermo Scientific Alfa Aesar 011122.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bismuth(III) nitrate oxide is used as an organic chemical synthesis intermediate. It is also used in the spectrophotometric detection of bismuth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-46-3
Molecular Formula BiNO4
Molecular Weight (g/mol) 286.98
MDL Number MFCD00064844
InChI Key OGCYGKRWXWQOAK-UHFFFAOYSA-N
Synonym Bismuth oxynitrate
IUPAC Name (nitrooxy)bismuthanone
SMILES [O-][N+](=O)O[Bi]=O

Specifications

Physical Form Powder
Quantity 50 g
UN Number UN1477
Solubility Information Insoluble in water. Soluble in acids.
Formula Weight 286.99
Percent Purity 99.998%
Grade Puratronic™
Assay (metals basis)
Chemical Name or Material Bismuth(III) nitrate oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ03K024 Certificate TypeCertificate of Analysis Date19/04/2024 Catalog Number011122.18,011122.09
Lot NumberS21J018 Certificate TypeCertificate of Analysis Date27/06/2023 Catalog Number011122.18,011122,011122.09
Lot NumberN28J056 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number011122.18,011122.09
Lot NumberS30I011 Certificate TypeCertificate of Analysis Date20/07/2022 Catalog Number011122.18,011122.09
Lot NumberX16G019 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011122.18,011122.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth(III) nitrate oxide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.