Learn More
Bismuth(III) nitrate oxide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Description
Bismuth(III) nitrate oxide is used as an organic chemical synthesis intermediate. It is also used in the spectrophotometric detection of bismuth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Powder |
Quantity | 10 g |
UN Number | UN1477 |
Solubility Information | Insoluble in water. Soluble in acids. |
Formula Weight | 286.99 |
Percent Purity | 99.998% |
Grade | Puratronic™ |
Assay | (metals basis) |
Chemical Name or Material | Bismuth(III) nitrate oxide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Q03K024 | Certificate of Analysis | 19/04/2024 | 011122.18,011122.09 |
S21J018 | Certificate of Analysis | 27/06/2023 | 011122.18,011122,011122.09 |
N28J056 | Certificate of Analysis | 09/05/2023 | 011122.18,011122.09 |
S30I011 | Certificate of Analysis | 20/07/2022 | 011122.18,011122.09 |
X16G019 | Certificate of Analysis | 12/04/2022 | 011122.18,011122.09 |
Safety and Handling


- Bismuth nitrate oxide
- Danger
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.