missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ BlockAid™ Blocking Solution

BlockAid™ Blocking Solution

Brand:  Molecular Probes™ B10710

120.08 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.


Product Code. 11514147

  • £163.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

BlockAid™ Blocking Solution is superior to commonly used blocking reagents such as bovine serum albumin, normal serum, or casein for use with cells or tissue sections. It is an optimized mix of protein-blocking components, with no dilution required. BlockAid™ Blocking Solution can be used as the initial blocking reagent before primary antibody is applied to minimize non-specific antibody binding, and as a diluent for both the primary and secondary antibody. This reagent is appropriate for cell or tissue immunofluorescence applications. It has also been validated for use with streptavidin conjugates and microsphere-protein conjugates.

Important Features of BlockAid™ Blocking Solution:

  • Excellent background reduction—superior to conventional blocking solutions

  • Ready to use—no dilution or stock preparation required

  • Versatile—use with any primary or secondary antibody, streptavidin conjugate, microsphere protein conjugates, or Qdot™ conjugates

    Minimizing background from non-specific protein binding of antibodies is essential for improved sensitivity and maximum signal-to-background ratios. This is especially crucial when looking for low-expressing antigens, when using samples with high autofluorescence, or for techniques where signals are inherently dim (such as use of directly-labeled primary antibodies or use of super-resolution imaging).
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Blocking Solution
    Blocking⁄Background Suppression Reagent
    50 mL
    BlockAid
    Store in freezer -5°C to -30°C.
    Wet Ice
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates

    Certificates

    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot Number3096435 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog NumberB10710
    Lot Number3096395 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog NumberB10710
    Lot Number3068197 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog NumberB10710
    Lot Number3008193 Certificate TypeCertificate of Analysis Date30/07/2024 Catalog NumberB10710
    Lot Number2845268 Certificate TypeCertificate of Analysis Date04/03/2024 Catalog NumberB10710
    Special Offers

    Special Offers

    • Multiple Manufacturers
      Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
      Promo Code: 24155

      Expires2025-04-30

      Days

      23

      Hours

      3

      Minutes

      33

      Seconds

      48
      Hot Deal

      *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

      Terms & Conditions See Full Promo Details
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Molecular Probes™ BlockAid™ Blocking Solution > 50mL

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.