missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borane-dimethyl sulfide complex, 2M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon

Specifications
Specifications
Color | Colorless to Yellow |
Density | 0.855 g/mL |
Flash Point | −17°C (−1°F) |
Odor | Characteristic |
Packaging | ChemSeal™ bottles |
Linear Formula | (CH3)2S·BH3 |
Quantity | 500 mL |
UN Number | UN3399 |
Beilstein | 3663489 |
Sensitivity | Air and moisture sensitive |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y17I006 | Certificate of Analysis | 24/10/2023 | 042965.AE,042965.AP |
M17J022 | Certificate of Analysis | 23/01/2023 | 042965.AE,042965.AP |
G29T046 | Certificate of Analysis | 08/12/2021 | 042965.AE,042965.AP |
K24N01 | Certificate of Analysis | 08/12/2021 | 042965.AE,042965.AP |
E07S027 | Certificate of Analysis | 08/12/2021 | 042965.AE,042965.AP |
Safety and Handling




Signal Word
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
- H225-Highly flammable liquid and vapour.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H360FD-May damage fertility. May damage the unborn child.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Borane-dimethyl sulfide complex, 2M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction