missing translation for 'onlineSavingsMsg'
Learn More
Boron triiodide, 95%, Thermo Scientific Chemicals Product Code.: 11374949

Boron triiodide, 95%, Thermo Scientific Chemicals

Product Code. 11374949
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11374949

Brand: Thermo Scientific Alfa Aesar 087220.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Boron triiodide is used as pharmaceutical, chemical and organic intermediate. It is also used as a catalyst and for preparation of boron compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13517-10-7
Molecular Formula BI3
Molecular Weight (g/mol) 391.52
MDL Number MFCD00036286
InChI Key YMEKEHSRPZAOGO-UHFFFAOYSA-N
Synonym boron triiodide, borane, triiodo, triiodoboron, acmc-1bo2g, boron triiodide 1g, inchi=1/bi3/c2-1 3
PubChem CID 83546
IUPAC Name triiodoborane
SMILES IB(I)I

Specifications

Boiling Point 210°C
Melting Point 48°C to 51°C
Color White
Physical Form Crystals
Quantity 1 g
Assay Percent Range >98%
Solubility Information Reacts with water.
Formula Weight 391.52
Odor Pungent
Sensitivity Light and moisture sensitive
Density 3.35
Chemical Name or Material Boron triiodide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX28K010 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number087220.06,087220.03,087220.14
Lot NumberS28K019 Certificate TypeCertificate of Analysis Date25/07/2024 Catalog Number087220.06,087220.03,087220.14
Lot NumberY16J024 Certificate TypeCertificate of Analysis Date15/12/2023 Catalog Number087220.06,087220.03,087220.14
Lot NumberR12J036 Certificate TypeCertificate of Analysis Date25/05/2023 Catalog Number087220.06,087220.03
Lot NumberP16I052 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number087220.06,087220.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boron triiodide, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.