Learn More
BRAND™ Single-Channel Micropipette Transferpette™ Electronic, DE-M
Ergonomic, handy design. Clear and intuitive operation. Up to 4,000 pipetting cycles on a single charge.
£292.25 - £524.00
Specifications
Autoclavable Autoclavable
Number of Channels 1
Product Line Transferpette
View More Specs
Products 11
Product Code Brand Volume (Metric) Color Includes Price Quantity & Availability  
Product Code Brand Volume (Metric) Color Includes Price Quantity & Availability  
10641603
View Documents
BRAND™
705339
0.5 to 10 μL Gray Without AC Adapter
£442.00
Each
10510882
View accessory products View Documents
BRAND™
705343
10 to 200 μL Yellow Without AC Adapter
£436.00
Each
10120352
View accessory products View Documents
BRAND™
705340
2 to 20 μL Gray Without AC Adapter
£442.00
Each
12398593
View Documents
BRAND™
705327
250 to 5000 μL Red USA/Japan AC Adapter N/A N/A N/A
10102472
View Documents
BRAND™
705347
250 to 5000 μL Red Without AC Adapter
£442.00
Each
10511072
View accessory products View Documents
BRAND™
705346
50 to 1000 μL Blue Without AC Adapter
£442.00
Each
12348593
View Documents
BRAND™
705317
500 to 5000 μL Red UK/Ireland AC Adapter
£475.00
Each
12338593
View accessory products View Documents
BRAND™
705316
100 to 1000 μL Blue UK/Ireland AC Adapter
£524.00
Each
12318593
View accessory products View Documents
BRAND™
705310
2 to 20 μL Gray UK/Ireland AC Adapter
£466.00
Each
12308593
View Documents
BRAND™
705309
0.5 to 10 μL UK/Ireland plug
£480.00
Each
12328593
View accessory products View Documents
BRAND™
705313
10 to 200 μL UK/Ireland plug
£524.00
Each
Description
Description
Transferpette™ electronic pipettes combine the globally proven properties of mechanical piston operated air displacement pipettes by BRAND with the advantages of an electronic device.
Features:
  • The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).
  • Five convenient programs: pipetting, reverse pipetting, mixing, GEL-Electrophoresis and dispensing.
  • Easy Calibration-Technique for making quick adjustments of the Transferpette™ electronic, without tools.
  • Limitations of use:
  • Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F) (other temperatures upon request)
  • Vapor pressure up to 500 mbar
  • Viscosity: 260 mPa s
  • For viscous media, the speed must be adjusted if necessary.
Specifications
Specifications
Autoclavable
1
Transferpette
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
BRAND™ Single-Channel Micropipette Transferpette™ Electronic, DE-M