Learn More
BTX™ HT 25-well Electroporation Plates
HT 25-well Electroporation Plates for use with ECM 630 and ECM 830 Electroporation Systems.
Brand: BTX™ 450467
Description
- 25-well disposable electroporation plate
- Standard multi-well formats
- Bio-compatible materials
Specifications
siRNA Transfection, Library Studies and Stem Cell Project | |
Medical grade polycarbonate, polished aluminium | |
Electroporation consumables | |
25-well Electroporation Plate | |
6 Pk. | |
2 mm | |
Sterile | |
Yes |
ECM 630HT, ECM830HT, Gemini X2 HT Electroporation System | |
BTX Electroporation | |
25-Well Disposable Electroporation Plate, 2 mm gap, 100 μl, pkg. of 6 | |
6 plates plus seals | |
0.078 in. | |
100 μL (Working) | |
Exponential Decay Wave, Square Wave |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.