Learn More
Macherey-Nagel™ Butyl Phase Polygosil™ C4
Brand:  Macherey-Nagel™ 711680.10
 View more versions of this product
Product Code. 10556291
  • £197.00 / 10g
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
7 μm
Liquid Chromatography
Polygosil C4 Phases
300 Å
10 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Butyl Phase Polygosil™ C4 > Quantity: 10g; Pore Size: 300 Angstroms