Learn More
Masterflex™ Transfer Tubing, C-Flex™, Opaque White
Meets the demands of various industries
£34.60 - £143.00
Specifications
Autoclavable | Autoclavable |
---|---|
Min. Operating Temperature (Metric) | -45°C |
Color | White |
Product Code | Brand | Diameter (Metric) Outer | Diameter (Metric) Inner | Maximum Pressure | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Diameter (Metric) Inner | Maximum Pressure | Price | Quantity & Availability | ||||
11717529
|
Masterflex™
06424-66 |
6.4 mm | 3 mm | 9 psi |
£48.40
7.50m |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
11783149
|
Masterflex™
06424-69 |
11.1 mm | 4.8 mm | 22 psi |
£143.00
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
11797509
|
Masterflex™
06424-60 |
2.4 mm | 4.8 mm | 31 psi |
£34.60
7.50m |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||
11757529
|
Masterflex™
06424-75 |
11.1 mm | 7.9 mm | 10 psi |
£80.70
7.50m |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
11767529
|
Masterflex™
06424-76 |
12.7 mm | 7.9 mm | 16 psi |
£104.00
7.50m |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||
11777529
|
Masterflex™
06424-12 |
12.7 mm | 9.6 mm | 10 psi |
£89.50
7.50m |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
11787529
|
Masterflex™
06424-79 |
15.9 mm | 9.6 mm | 14 psi |
£140.00
7.50m |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
11787509
|
Masterflex™
06424-59 |
2.1 mm | 0.51 mm | 2 bar |
£44.50
7.50m |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||||
11727529
|
Masterflex™
06424-71 |
9.6 mm | 6.4 mm | 12 psi |
£103.00
7.50m |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||
11707519
|
Masterflex™
06424-62 |
3.2 mm | 1.6 mm | 1 bar |
£43.60
7.50m |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
11797519
|
Masterflex™
06424-67 |
6.4 mm | 3 mm | 21 psi |
£47.70
7.50m |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||
11747529
|
Masterflex™
06424-74 |
12.7 mm | 6.4 mm | 17 psi |
£135.00
7.50m |
Estimated Shipment: 23-07-2025
Log in to see stock. |
||||
Description
Meets the demands of various industries
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Regulatory approvals to match your unique application
Specifications
Autoclavable | |
-45°C | |
White |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Masterflex™ Transfer Tubing, C-Flex™, Opaque White