missing translation for 'onlineSavingsMsg'
Learn More
Calcium bromide hydrate, for analysis, Thermo Scientific Chemicals Product Code.: 10480911

Calcium bromide hydrate, for analysis, Thermo Scientific Chemicals

Product Code. 10480911
500 g, Plastic bottle
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10480911

Brand: Thermo Scientific Acros 300845000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71626-99-8
Molecular Formula Br2Ca
Molecular Weight (g/mol) 199.89
MDL Number MFCD00149608
InChI Key WGEFECGEFUFIQW-UHFFFAOYSA-L
IUPAC Name calcium dibromide
SMILES [Ca++].[Br-].[Br-]

Specifications

Melting Point 730.0°C
Color White
Physical Form Crystalline Powder and/or Chunks
Quantity 500 g
Linear Formula CaBr2·xH2O
Solubility Information Solubility in water: 1420g/L (20°C). Other solubilities: soluble in methanol, ethanol
Formula Weight 199.9
Percent Purity ≥83.0% (Complexometry) (CaBr2)
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Calcium bromide hydrate, For analysis
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465243 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog Number300845000,300840010,300840025
Lot NumberA0452652 Certificate TypeCertificate of Analysis Date04/05/2023 Catalog Number300845000,300840010,300840025
Lot NumberA0451656 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number300845000,300840010,300840025
Lot NumberA0451065 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number300845000,300840010,300840025
Lot NumberA0450285 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number300845000,300840010,300840025
compliance-icons
Product Identifier
  • Calcium bromide hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Calcium bromide, hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium bromide hydrate, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.