missing translation for 'onlineSavingsMsg'
Learn More
Calcium L-threonate, 98%, Thermo Scientific Chemicals Product Code.: 15447579

Calcium L-threonate, 98%, Thermo Scientific Chemicals

Product Code. 15447579
100 g
Click to view available options
Quantity:
100 g
250 g
500 g
This item is not returnable. View return policy

Product Code. 15447579

Brand: Thermo Scientific Alfa Aesar J66279.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.

Solubility
Soluble in water. Sparingly soluble in methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70753-61-6
Molecular Formula C8H14CaO10
Molecular Weight (g/mol) 310.27
MDL Number MFCD00077882,MFCD00150824
InChI Key ZJXGOFZGZFVRHK-UHFFFAOYNA-L
Synonym calcium l-threonate, l-threonic acid calcium salt, calcium 2r,3s-2,3,4-trihydroxybutanoate, calcium threonate, unii-hbb4ypo93u, hbb4ypo93u, calcium bis l-threonate, threonine, calcium salt, bis l-threonic acid calcium salt, butanoic acid, 2,3,4-trihydroxy-, calcium salt 2:1 , 2r,3sShow MoreShow Less
PubChem CID 13388558
IUPAC Name calcium;(2R,3S)-2,3,4-trihydroxybutanoate
SMILES [Ca++].OCC(O)C(O)C([O-])=O.OCC(O)C(O)C([O-])=O

Specifications

Melting Point >300°C
Color Brown
Quantity 100 g
Solubility Information Soluble in water. Sparingly soluble in methanol.
Formula Weight 310.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Calcium L-threonate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP14J007 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberJ66279.22,J66279.36
Lot NumberT01I001 Certificate TypeCertificate of Analysis Date12/07/2022 Catalog NumberJ66279.22,J66279.36
Lot NumberQ11C072 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ66279.22,J66279.36
Lot NumberR26G060 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ66279.22,J66279.36
Lot NumberY23B020 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ66279.22,J66279.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium L-threonate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.