missing translation for 'onlineSavingsMsg'
Learn More
(+)-Camptothecin, 98%, Thermo Scientific Chemicals Product Code.: 10502461

(+)-Camptothecin, 98%, Thermo Scientific Chemicals

Product Code. 10502461
100 mg, Glass bottle
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10502461

Brand: Thermo Scientific Acros 276721000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7689-03-4
Molecular Formula C20H16N2O4
Molecular Weight (g/mol) 348.35
MDL Number MFCD00081076
InChI Key VSJKWCGYPAHWDS-FQEVSTJZSA-N
Synonym camptothecin, camptothecine, s-+-camptothecin, campathecin, +-camptothecine, d-camptothecin, 20 s-camptothecine, +-camptothecin, 21,22-secocamptothecin-21-oic acid lactone, s-camptothecinShow MoreShow Less
PubChem CID 24360
ChEBI CHEBI:27656
SMILES CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 260.0°C
Color Yellow
Loss on Drying 1.0% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Merck Index 15, 1737
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,acetonirile,dmso,,chloroform,hydrochloric acid
Formula Weight 348.35
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (+)-Camptothecin, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464842 Certificate TypeCertificate of Analysis Date12/08/2024 Catalog Number276720010,276721000
Lot NumberA0464834 Certificate TypeCertificate of Analysis Date08/08/2024 Catalog Number276720010,276721000
Lot NumberA0447855 Certificate TypeCertificate of Analysis Date06/12/2023 Catalog Number276720010,276721000
Lot NumberA0419913 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog Number276720010,276721000
Lot NumberA0406186 Certificate TypeCertificate of Analysis Date22/03/2021 Catalog Number276720010,276721000
compliance-icons
Product Identifier
  • (+)-Camptothecin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(+)-Camptothecin, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.