missing translation for 'onlineSavingsMsg'
Learn More
Cefixime, 98%, Thermo Scientific Chemicals Product Code.: 15589473

Cefixime, 98%, Thermo Scientific Chemicals

Product Code. 15589473
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15589473

Brand: Thermo Scientific Acros 459480010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79350-37-1
Molecular Formula C16H15N5O7S2
Molecular Weight (g/mol) 453.44
MDL Number MFCD00865020
InChI Key OKBVVJOGVLARMR-QSWIMTSFSA-N
Synonym cefixime, cefixima, cefiximum, cefixim, cefspan, cephoral, cefixoral, cefiximum latin, suprax, necopen
PubChem CID 5362065
ChEBI CHEBI:472657
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(carboxymethoxyimino)acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acidShow MoreShow Less
SMILES NC1=NC(=CS1)C(=N\OCC(O)=O)\C(=O)N[C@H]1[C@H]2SCC(C=C)=C(N2C1=O)C(O)=O

Specifications

pH 2.6 to 4.1
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 453.45
Percent Purity 98%
Chemical Name or Material Cefixime

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0407776 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number459480010,459480050
Lot NumberA0452134 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number459480010,459480050
Lot NumberA0445572 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number459480010,459480050
Lot NumberA0397125 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number459480010,459480050
Lot NumberA0399022 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number459480010,459480050
Product Identifier
  • Cefixime

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cefixime, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.