missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Cell Fractionation Buffer for PrepSEQ™ Nucleic Acid Sample Preparation Kit

Cell Fractionation Buffer is used to process the mammalian cell pellet to obtain free Mycoplasma and then combine with the Mycoplasma pellet in the large scale PrepSEQ extraction protocol.

Brand:  Applied Biosystems™ 4403461

Product Code. 30000676

  • £76.75 / 25mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Cell Fractionation Buffer is used to process the mammalian cell pellet to obtain free Mycoplasma and then combine with the Mycoplasma pellet in the large scale PrepSEQ extraction protocol. This protocol can process up to 15 mL of sample (up to 106 cells/mL) for the detection of Mycoplasma.

Specifications

Specifications

PrepSEQ™ Nucleic Acid Sample Preparation Kit
Sufficient for 15 mL of sample (up to 106 cells/mL)

Store at 2°C to 8°C.
Mycoplasma detection
3 bottles, 25 mL
Liquid
Cell Fractionation Buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2820363 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4403461
Lot Number2854274 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number4403461
Lot Number2980964 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number4403461
Lot Number3024524 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number4403461
Lot Number3053683 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number4403461
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ Cell Fractionation Buffer for PrepSEQ™ Nucleic Acid Sample Preparation Kit > Quantity: 25 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.