Learn More
Cesium acetate, 99% (metals basis), Thermo Scientific Chemicals
Description
Cesium acetate is used in organic synthesis especially in Perkin reaction to prepare unsaturated cinnamic-type acids. It is used in petroleum drilling fluids as an alternative to cesium formate. In organometallic chemistry, it is an additive for metal-ctalyzed coupling reactions. It is also used in the inversion of stereo chemistry of secondary alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 194°C |
Color | White |
Physical Form | Lump/Powder |
Quantity | 50 g |
Assay Percent Range | 99% (Metal basis) |
Solubility Information | Soluble in water and dimethyl sulfoxide. |
Formula Weight | 191.95 |
Appearance | Off-white crystalline powder |
Chemical Name or Material | Cesium acetate |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W07I006 | Certificate of Analysis | 15/09/2022 | 043095.30,043095.18 |
U11H079 | Certificate of Analysis | 30/05/2022 | 043095.30,043095.18 |
N10I002 | Certificate of Analysis | 12/04/2022 | 043095.30,043095.18 |
F17N20 | Certificate of Analysis | 18/11/2021 | 043095.30,043095.18 |
T24B042 | Certificate of Analysis | 18/11/2021 | 043095.30,043095.18 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.