missing translation for 'onlineSavingsMsg'
Learn More
Cesium acetate, 99% (metals basis), Thermo Scientific Chemicals Product Code.: 11344758

Cesium acetate, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11344758
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11344758

Brand: Thermo Scientific Alfa Aesar 043095.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cesium acetate is used in organic synthesis especially in Perkin reaction to prepare unsaturated cinnamic-type acids. It is used in petroleum drilling fluids as an alternative to cesium formate. In organometallic chemistry, it is an additive for metal-ctalyzed coupling reactions. It is also used in the inversion of stereo chemistry of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3396-11-0
Molecular Formula C2H3CsO2
Molecular Weight (g/mol) 191.949
MDL Number MFCD00013056
InChI Key ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
Synonym cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ionShow MoreShow Less
PubChem CID 5152919
IUPAC Name cesium;acetate
SMILES CC(=O)[O-].[Cs+]

Specifications

Melting Point 194°C
Color White
Physical Form Lump/Powder
Quantity 250 g
Assay Percent Range 99% (Metal basis)
Solubility Information Soluble in water and dimethyl sulfoxide.
Formula Weight 191.95
Appearance Off-white crystalline powder
Chemical Name or Material Cesium acetate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN27J036 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number043095.30
Lot NumberW07I006 Certificate TypeCertificate of Analysis Date15/09/2022 Catalog Number043095.30,043095.18
Lot NumberU11H079 Certificate TypeCertificate of Analysis Date30/05/2022 Catalog Number043095.30,043095.18
Lot NumberN10I002 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043095.30,043095.18
Lot NumberF17N20 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043095.30,043095.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium acetate, 99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.