missing translation for 'onlineSavingsMsg'
Learn More
Cesium bromide, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11302018

Cesium bromide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11302018
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11302018

Brand: Thermo Scientific Alfa Aesar 012928.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cesium bromide is applied to make radiation detectors and other measuring devices. Cesium bromide has limited application in the deep IR. Cesium bromide is slightly more amenable to optical working than CsI and is sometimes utilized as a beam splitter component in wideband spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-69-1
Molecular Formula BrCs
Molecular Weight (g/mol) 212.81
MDL Number MFCD00010954
InChI Key LYQFWZFBNBDLEO-UHFFFAOYSA-M
Synonym cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromideShow MoreShow Less
PubChem CID 24592
IUPAC Name cesium;bromide
SMILES [Br-].[Cs+]

Specifications

Boiling Point 1300°C
Melting Point 636°C
Color White
Physical Form Crystalline
Quantity 250 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information Soluble in water.
Formula Weight 212.81
Odor Odorless
Refractive Index 1.6984
Sensitivity Hygroscopic
Density 4.44
Chemical Name or Material Cesium bromide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU06K025 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog Number012928.18,012928.30
Lot Number10251927 Certificate TypeCertificate of Analysis Date02/08/2024 Catalog Number012928.18,012928.30
Lot NumberS05K057 Certificate TypeCertificate of Analysis Date25/06/2024 Catalog Number012928.18,012928.30
Lot NumberM20I076 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012928.18,012928.30
Lot NumberJ11L22 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number012928.18,012928.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium bromide, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.