missing translation for 'onlineSavingsMsg'
Learn More
Cesium carbonate, 99.5%, for analysis, Thermo Scientific Chemicals Product Code.: 11984311

Cesium carbonate, 99.5%, for analysis, Thermo Scientific Chemicals

Product Code. 11984311
2.5 kg, Plastic bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 11984311

Brand: Thermo Scientific Acros 192040025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-17-8
Molecular Formula CCs2O3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00010957
InChI Key FJDQFPXHSGXQBY-UHFFFAOYSA-L
Synonym cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonateShow MoreShow Less
PubChem CID 10796
SMILES [Cs+].[Cs+].[O-]C([O-])=O

Specifications

Melting Point >400°C
Color White
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 99.4% min. (By difference)
Linear Formula Cs2CO3
Fieser 07,57; 08,81; 09,99; 12,108; 13,70; 14,77; 15,73
Merck Index 15,2009
Solubility Information Solubility in water: 261 g/100mL water (20°C). Other solubilities: soluble in alcohol and ether
IUPAC Name dicaesium(1+) carbonate
Formula Weight 325.82
Percent Purity 99.5%
Grade Analytical
Loss on Drying 0.5% max. (500°C)
Packaging Plastic bottle
Specific Gravity 4.072
Density 4.0720g/mL
Chemical Name or Material Cesium carbonate, 99.5%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458837 Certificate TypeCertificate of Analysis Date15/12/2023 Catalog Number192040250,192040050,192045000,192041000,192040025
Lot NumberA0456482 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number192040250,192040050,192045000,192041000,192040025
Lot NumberA0454851 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number192040250,192040050,192045000,192041000,192040025
Lot NumberA0451600 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number192040250,192040050,192045000,192041000,192040025
Lot NumberA0400185 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number192040250,192040050,192045000,192041000,192040025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cesium carbonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P263-Avoid contact during pregnancy/while nursing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium carbonate, 99.5%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.